ENAH5802EBF0 Enamine

ethyl 3-(2-chloro-4-methoxyphenyl)-3-oxopropanoate

  • MDL number MFCD12911835
  • Cas Number : 941696-11-3
  • Molecular Weight : 256.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-(2-chloro-4-methoxyphenyl)-3-oxopropanoate
Inchi Code 1S/C12H13ClO4/c1-3-17-12(15)7-11(14)9-5-4-8(16-2)6-10(9)13/h4-6H,3,7H2,1-2H3
Inchi Key OYFMBFWBWVAOGI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7424807

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7424807
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart