ENAH5802EBFD Enamine

2-tert-butyl 4-methyl 5-oxo-octahydrocyclopenta[c]pyrrole-2,4-dicarboxylate

  • MDL number MFCD23102233
  • Cas Number : 1369496-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7425037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7425037
Availability details for
Quantity
  • Availability
  • Add to Cart