ENAH5802EC01 Enamine

2-(2-chlorophenyl)-2-(methylamino)-6-(phenylmethylidene)cyclohexan-1-one

  • Cas Number : 790272-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02738
Availability details for
Quantity
  • Availability
  • Add to Cart