ENAH5802EC17 Enamine

(6R)-2,2,6-trimethylmorpholine hydrochloride

  • MDL number MFCD32198466
  • Cas Number : 2375248-66-9
  • Molecular Weight : 165.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7443689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443689
Availability details for
Quantity
  • Availability
  • Add to Cart