ENAH5802EC18 Enamine

(6S)-2,2,6-trimethylmorpholine hydrochloride

  • Cas Number : 2375247-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7442956

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442956
Availability details for
Quantity
  • Availability
  • Add to Cart