ENAH5802EC19 Enamine

3-[(dimethylamino)methyl]oxetan-3-amine

  • MDL number MFCD30619810

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-4842031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4842031
Availability details for
Quantity
  • Availability
  • Add to Cart