ENAH5802EC1A Enamine

3-(dimethylphosphoryl)propanenitrile

  • Cas Number : 53314-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7442273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442273
Availability details for
Quantity
  • Availability
  • Add to Cart