ENAH5802EC1B Enamine

N-propylmethanesulfonoimidamide hydrochloride

  • MDL number MFCD32066594
  • Cas Number : 2305948-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432738
Availability details for
Quantity
  • Availability
  • Add to Cart