ENAH5802EC1C Enamine

3-(difluoromethyl)morpholine hydrochloride

  • MDL number MFCD32069233
  • Cas Number : 2375261-03-1
  • Molecular Weight : 173.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(difluoromethyl)morpholine hydrochloride
Inchi Code 1S/C5H9F2NO.ClH/c6-5(7)4-3-9-2-1-8-4;/h4-5,8H,1-3H2;1H
Inchi Key ZLWGWWNQUDAVLR-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7438975

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7438975
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart