ENAH5802EC1E Enamine

2-cyclobutylmorpholine hydrochloride

  • MDL number MFCD28893096

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-7434462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434462
Availability details for
Quantity
  • Availability
  • Add to Cart