ENAH5802EC26 Enamine

4,4-dimethyl-1,4-azasilepane

  • MDL number MFCD32198261
  • Cas Number : 1330276-96-4
  • Molecular Weight : 143.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,4-dimethyl-1,4-azasilepane
Inchi Code 1S/C7H17NSi/c1-9(2)6-3-4-8-5-7-9/h8H,3-7H2,1-2H3
Inchi Key KWEOZTVWBFWMKC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6748268

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6748268
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart