ENAH5802EC28 Enamine

3-(methoxymethyl)-3-methylmorpholine

  • MDL number MFCD30095706
  • Cas Number : 777838-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-2971730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2971730
Availability details for
Quantity
  • Availability
  • Add to Cart