ENAH5802EC2D Enamine

3-[(tert-butoxy)carbonyl]-9-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-azabicyclo[3.3.1]nonane-7-carboxylic acid

  • MDL number MFCD32066403
  • Cas Number : 2309468-79-7
  • Molecular Weight : 506.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 9-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(tert-butoxycarbonyl)-3-azabicyclo[3.3.1]nonane-7-carboxylic acid
Inchi Code 1S/C29H34N2O6/c1-29(2,3)37-28(35)31-14-18-12-17(26(32)33)13-19(15-31)25(18)30-27(34)36-16-24-22-10-6-4-8-20(22)21-9-5-7-11-23(21)24/h4-11,17-19,24-25H,12-16H2,1-3H3,(H,30,34)(H,32,33)
Inchi Key JZIFUGFWDWEFGW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-183597

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-183597
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart