ENAH5802EC2E Enamine

2-(furan-3-yl)morpholine; oxalic acid

  • Molecular Weight : 243.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(furan-3-yl)morpholine oxalate

Description

Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427957
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart