ENAH5802EC33 Enamine

octahydro-1H-isoindole-5,6-diol hydrochloride

  • MDL number MFCD22570181
  • Cas Number : 2206824-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355477
Availability details for
Quantity
  • Availability
  • Add to Cart