ENAH5802EC34 Enamine

1-imino-1-oxo-1lambda6-thiane-4-carbonitrile

  • MDL number MFCD32069170
  • Cas Number : 2232916-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748558
Availability details for
Quantity
  • Availability
  • Add to Cart