ENAH5802EC35 Enamine

imino(methyl)(1-methyl-1H-imidazol-5-yl)-lambda6-sulfanone

  • Cas Number : 2375270-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6744014

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6744014
Availability details for
Quantity
  • Availability
  • Add to Cart