ENAH5802EC36 Enamine

imino(methyl)(1-methyl-1H-pyrazol-5-yl)-lambda6-sulfanone

  • MDL number MFCD32198182
  • Cas Number : 2375273-89-3
  • Molecular Weight : 159.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-methyl-S-methylene-1H-pyrazole-5-sulfinamide

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6744011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6744011
Availability details for
Quantity
  • Availability
  • Add to Cart