ENAH5802EC37 Enamine

(6S)-6-(methoxymethyl)-2,2-dimethylmorpholine

  • Cas Number : 2375248-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443107

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443107
Availability details for
Quantity
  • Availability
  • Add to Cart