ENAH5802EC38 Enamine

(6R)-6-(methoxymethyl)-2,2-dimethylmorpholine

  • MDL number MFCD32197982
  • Cas Number : 2375250-51-2
  • Molecular Weight : 159.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-6-(methoxymethyl)-2,2-dimethylmorpholine
Inchi Code 1S/C8H17NO2/c1-8(2)6-9-4-7(11-8)5-10-3/h7,9H,4-6H2,1-3H3/t7-/m1/s1
Inchi Key MVQWSVQLLPTBPV-SSDOTTSWSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7442894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442894
Availability details for
Quantity
  • Availability
  • Add to Cart