ENAH5802EC39 Enamine

3lambda4-thia-7-azabicyclo[3.3.1]nonan-3-one hydrochloride

  • Cas Number : 2375274-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443813
Availability details for
Quantity
  • Availability
  • Add to Cart