ENAH5802EC3A Enamine

2,2-difluorocyclopropane-1-sulfonyl fluoride

  • Cas Number : 1936720-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-245261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-245261
Availability details for
Quantity
  • Availability
  • Add to Cart