ENAH5802EC40 Enamine

[3-(dimethylamino)propyl](imino)methyl-lambda6-sulfanone dihydrochloride

  • Molecular Weight : 237.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(dimethylamino)-S-methylpropane-1-sulfinamide dihydrochloride
Inchi Code 1S/C6H16N2OS.2ClH/c1-8(2)5-4-6-10(3,7)9;;/h6H,4-5H2,1-3H3,(H2,7,9);2*1H
Inchi Key WUTOOQGKLKZGJM-UHFFFAOYSA-N

Description

Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7433717

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433717
Availability details for
Quantity
  • Availability
  • Add to Cart