ENAH5802EC42 Enamine

(dimethyl-S-aminosulfonimidoyl)(propyl)amine hydrochloride

  • MDL number MFCD32066593
  • Cas Number : 2305949-05-5
  • Molecular Weight : 201.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C5H15N3OS.ClH/c1-4-5-7-10(6,9)8(2)3;/h4-5H2,1-3H3,(H2,6,7,9);1H
Inchi Key PGQOSZAZFLSDED-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432735
Availability details for
Quantity
  • Availability
  • Add to Cart