ENAH5802EC45 Enamine

N-phenylmethanesulfonoimidamide hydrochloride

  • MDL number MFCD31924650
  • Cas Number : 2305948-99-4
  • Molecular Weight : 206.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C7H10N2OS.ClH/c1-11(8,10)9-7-5-3-2-4-6-7;/h2-6H,1H2,(H3,8,9,10);1H
Inchi Key KZYCTLYIWNTVGH-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7356770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356770
Availability details for
Quantity
  • Availability
  • Add to Cart