ENAH5802EC48 Enamine

1-(dimethylphosphoryl)-3-fluorobenzene

  • MDL number MFCD32068704
  • Cas Number : 23588-14-9
  • Molecular Weight : 172.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-fluorophenyl)dimethylphosphine oxide
Inchi Code 1S/C8H10FOP/c1-11(2,10)8-5-3-4-7(9)6-8/h3-6H,1-2H3
Inchi Key IQSFNVZHXDRRFG-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6760258

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6760258
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart