ENAH5802EC4A Enamine

3H,4H,6H,7H,8H-2lambda6-pyrrolo[2,1-c][1,2,4,6]thiatriazine-2,2-dione

  • MDL number MFCD32068805
  • Cas Number : 1254099-88-1
  • Molecular Weight : 175.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 178-180
Iupac Name 3,4,6,7-tetrahydro-1H-pyrrolo[2,1-c][1,2,4,6]thiatriazine 2,2-dioxide
Inchi Code 1S/C5H9N3O2S/c9-11(10)6-4-8-3-1-2-5(8)7-11/h2,6-7H,1,3-4H2
Inchi Key ZSPYGEQVYMUCPF-UHFFFAOYSA-N

Description

Melting Point 178-180
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1721340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721340
Availability details for
Quantity
  • Availability
  • Add to Cart