ENAH5802EC4B Enamine

(R)-cyclopentyl(dimethylphosphoryl)methanamine hydrochloride

  • MDL number MFCD31924372
  • Cas Number : 2343964-33-8
  • Molecular Weight : 211.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7355400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355400
Availability details for
Quantity
  • Availability
  • Add to Cart