ENAH5802EC4C Enamine

(S)-cyclopentyl(dimethylphosphoryl)methanamine

  • MDL number MFCD31924310
  • Cas Number : 2343963-93-7
  • Molecular Weight : 175.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7353974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353974
Availability details for
Quantity
  • Availability
  • Add to Cart