ENAH5802EC4E Enamine

2lambda6-thia-3-azaspiro[4.4]nonane-2,2-dione

  • Cas Number : 2344679-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7424697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7424697
Availability details for
Quantity
  • Availability
  • Add to Cart