ENAH5802EC51 Enamine

5-hydroxypyridine-2-sulfonyl fluoride

  • MDL number MFCD32065946
  • Cas Number : 2361644-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355309
Availability details for
Quantity
  • Availability
  • Add to Cart