ENAH5802EC65 Enamine

4-(3,3-difluorooxetan-2-yl)benzonitrile

  • MDL number MFCD32066563
  • Cas Number : 2361645-20-5
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6739528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739528
Availability details for
Quantity
  • Availability
  • Add to Cart