ENAH5802ECD6 Enamine

4-amino-1,3-dimethyl-1H-pyrazole-5-carbonitrile

  • MDL number MFCD18431086
  • Cas Number : 32183-14-5
  • Molecular Weight : 136.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-189878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189878
Availability details for
Quantity
  • Availability
  • Add to Cart