ENAH5802ECF9 Enamine

2,6-dimethyl-4-sulfanylidene-1,4-dihydropyrimidine-5-carbonitrile

  • MDL number MFCD23146430
  • Cas Number : 13996-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-212
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27353
Availability details for
Quantity
  • Availability
  • Add to Cart