ENAH5802ED10 Enamine

5-nitronaphthalene-1-carbonitrile

  • MDL number MFCD01712220
  • Cas Number : 23245-64-9
  • Molecular Weight : 198.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7435337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435337
Availability details for
Quantity
  • Availability
  • Add to Cart