ENAH5802ED3A Enamine

tert-butyl 3-[2-(acetylsulfanyl)ethyl]-3-cyanoazetidine-1-carboxylate

  • MDL number MFCD32066557
  • Cas Number : 2361645-17-0
  • Molecular Weight : 284.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6733540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6733540
Availability details for
Quantity
  • Availability
  • Add to Cart