ENAH5802ED73 Enamine

2,4,5-trimethyl-2,3-dihydro-1H-pyrazol-3-imine

  • MDL number MFCD26143820
  • Cas Number : 1365633-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71463
Availability details for
Quantity
  • Availability
  • Add to Cart