ENAH5802EDB2 Enamine

(3aS,7aS)-octahydro-1,3-benzoxazol-2-one

  • MDL number MFCD19217115
  • Cas Number : 189942-51-6
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6763966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763966
Availability details for
Quantity
  • Availability
  • Add to Cart