ENAH5802EDBB Enamine

3-propyl-2,5-dihydro-1,2,4-thiadiazol-5-imine hydrochloride

  • MDL number MFCD28656855
  • Cas Number : 1803560-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199456

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199456
Availability details for
Quantity
  • Availability
  • Add to Cart