ENAH5802EE21 Enamine

2-hydroxypropane-1,2,3-tricarboxylic acid; 4-[4-(3-methylphenyl)-5-({[3-(2-methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}sulfanyl)-4H-1,2,4-triazol-3-yl]morpholine

  • MDL number MFCD32069098
  • Cas Number : 2375260-23-2
  • Molecular Weight : 640.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data C6H8O7
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7437995

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7437995
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart