ENAH5802EE53 Enamine

6-chloro-1-methyl-1H,2H,4H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD28155386
  • Cas Number : 1934816-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62624
Availability details for
Quantity
  • Availability
  • Add to Cart