ENAH5802EE78 Enamine

5-(2,2,2-trifluoroethyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD32068641
  • Cas Number : 2361683-73-8
  • Molecular Weight : 177.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7427719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427719
Availability details for
Quantity
  • Availability
  • Add to Cart