ENAH5802EE7D Enamine

2-[(5-methylfuran-2-yl)methyl]-2,3-dihydro-1H-pyrazol-3-imine hydrochloride

  • MDL number MFCD23145021
  • Cas Number : 1955505-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-65975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65975
Availability details for
Quantity
  • Availability
  • Add to Cart