ENAH5802EE84 Enamine

4-(piperidin-1-yl)-1,2-dihydropyridin-2-one

  • MDL number MFCD28338080
  • Cas Number : 1006052-99-8
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426926
Availability details for
Quantity
  • Availability
  • Add to Cart