ENAH5802EE88 Enamine

2,5-dimethyl-4-(trifluoromethyl)-2,3-dihydro-1H-pyrazol-3-imine

  • MDL number MFCD29960034
  • Cas Number : 1934458-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207110
Availability details for
Quantity
  • Availability
  • Add to Cart