ENAH5802EE8D Enamine

N-cyclopropylbenzenecarbonimidoyl chloride

  • Cas Number : 869472-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14633
Availability details for
Quantity
  • Availability
  • Add to Cart