ENAH5802EE91 Enamine

4-[amino(nitroso)methylidene]-N,N-dimethyl-1,4-dihydropyridin-2-amine

  • Cas Number : 1955564-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76296
Availability details for
Quantity
  • Availability
  • Add to Cart