ENAH5802EE92 Enamine

diazenyl[2-(dimethylamino)-1,4-dihydropyridin-4-ylidene]methanol

  • Cas Number : 1937346-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73376
Availability details for
Quantity
  • Availability
  • Add to Cart