ENAH5802EE9B Enamine

N-(propan-2-yl)benzenecarbonimidoyl chloride

  • Cas Number : 6620-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14694
Availability details for
Quantity
  • Availability
  • Add to Cart