ENAH5802EEA5 Enamine

2H,3H,4H-pyridazino[4,5-b]indol-4-one

  • MDL number MFCD26143741
  • Cas Number : 1934916-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 326-328
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28227
Availability details for
Quantity
  • Availability
  • Add to Cart